Search
Search
| Gene Symbol: WASF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018056581.1 | XP_017912070.1 | 5 - 6 | 580 | 75 | |
| XM_018056582.1 | XP_017912071.1 | 5 - 6 | 651 | 75 | ||
| XM_018056583.1 | XP_017912072.1 | 5 - 6 | 580 | 75 | ||
| GenBank mRNA | JO352780.2 | - | 3 - 4 | 556 | 75 | |
| JO413199.2 | - | 3 - 4 | 560 | 75 | ||
Set Membership: |
|