Search
Search

MINDY1| Gene Symbol: MINDY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005677639.3 | XP_005677696.2 | 5 - 6 | 1690 | 56 | |
| XM_005677640.2 | XP_005677697.2 | 5 - 6 | 1668 | 56 | ||
| XM_005677641.3 | XP_005677698.2 | 5 - 6 | 1690 | 56 | ||
| XM_013962427.2 | XP_013817881.2 | 5 - 6 | 1690 | 56 | ||
| GenBank mRNA | KA313625.1 | - | 5 - 6 | 1372 | 56 | |
Set Membership: |
|