Search
Search

C1H3orf52| Gene Symbol: C1H3orf52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065255.1 | XP_017920744.1 | 1 - 2 | 236 | 72 | |
| XM_018065321.1 | XP_017920810.1 | 1 - 2 | 236 | 72 | ||
| GenBank mRNA | JO587315.2 | - | 1 - 2 | 208 | 72 | |
| JO587316.2 | - | 1 - 2 | 208 | 72 | ||
Set Membership: |
|