Search
Search
| Gene Symbol: LTN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005674720.3 | XP_005674777.2 | 17 - 18 | 3381 | 72 | |
| GenBank mRNA | JO366877.2 | - | 2 - 3 | 184 | 72 | |
| JO389490.2 | - | 3 - 4 | 471 | 72 | ||
| JO454504.2 | - | 3 - 4 | 316 | 72 | ||
| JO522698.2 | - | 3 - 4 | 405 | 72 | ||
| JT705976.1 | - | 17 - 18 | 3255 | 72 | ||
| JT717201.1 | - | 5 - 6 | 863 | 72 | ||
| JT738021.1 | - | 3 - 4 | 480 | 72 | ||
| KA330462.1 | - | 1 - 2 | 108 | 72 | ||
Set Membership: |
|