Search
Search

TSPAN19| Gene Symbol: TSPAN19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005679776.2 | XP_005679833.2 | 7 - 8 | 676 | 112 | |
| XM_018049084.1 | XP_017904573.1 | 6 - 7 | 599 | 112 | ||
Set Membership: |
|