Search
Search

DIAPH2| Gene Symbol: DIAPH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018044114.1 | XP_017899603.1 | 20 - 21 | 3247 | 90 | |
| GenBank mRNA | JO435123.2 | - | 3 - 4 | 994 | 90 | |
| JO593539.2 | - | 3 - 4 | 554 | 90 | ||
| JO593540.2 | - | 3 - 4 | 554 | 90 | ||
Set Membership: |
|