Search
Search
| Gene Symbol: WDR17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018042090.1 | XP_017897579.1 | 15 - 16 | 2285 | 60 | |
| XM_018042091.1 | XP_017897580.1 | 15 - 16 | 2284 | 60 | ||
| XM_018042092.1 | XP_017897581.1 | 14 - 15 | 2163 | 60 | ||
| XM_018042093.1 | XP_017897582.1 | 15 - 16 | 2293 | 60 | ||
Set Membership: |
|