Search
Search

KIAA0556| Gene Symbol: KIAA0556 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040845.1 | XP_017896334.1 | 16 - 17 | 2788 | 112 | |
| XM_018040846.1 | XP_017896335.1 | 15 - 16 | 2659 | 112 | ||
Set Membership: |
|