Search
Search

MAP7D3| Gene Symbol: MAP7D3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018044250.1 | XP_017899739.1 | 13 - 14 | 3029 | 110 | |
| GenBank mRNA | JO354657.2 | - | 4 - 5 | 719 | 110 | |
Set Membership: |
|