Search
Search

SLC2A8| Gene Symbol: SLC2A8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055805.1 | XP_017911294.1 | 7 - 8 | 1106 | 55 | |
| XM_018055806.1 | XP_017911295.1 | 6 - 7 | 994 | 55 | ||
| XM_018055807.1 | XP_017911296.1 | 7 - 8 | 1118 | 55 | ||
| GenBank mRNA | JO588366.2 | - | 3 - 4 | 923 | 55 | |
| JO630463.2 | - | 7 - 8 | 1100 | 55 | ||
Set Membership: |
|