Search
Search

SHROOM3| Gene Symbol: SHROOM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018049607.1 | XP_017905096.1 | 8 - 9 | 5397 | 64 | |
| XM_018049609.1 | XP_017905098.1 | 7 - 8 | 4969 | 64 | ||
| XM_018049610.1 | XP_017905099.1 | 7 - 8 | 5083 | 64 | ||
| XM_018049611.1 | XP_017905100.1 | 7 - 8 | 4918 | 64 | ||
Set Membership: |
|