Search
Search

LPCAT1| Gene Symbol: LPCAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065659.1 | XP_017921148.1 | 4 - 5 | 639 | 63 | |
| XM_018065660.1 | XP_017921149.1 | 4 - 5 | 882 | 63 | ||
Set Membership: |
|