Search
Search
| Gene Symbol: KIF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018066271.1 | XP_017921760.1 | 8 - 9 | 2109 | 56 | |
| XM_018066272.1 | XP_017921761.1 | 8 - 9 | 2472 | 56 | ||
| XM_018066273.1 | XP_017921762.1 | 8 - 9 | 2106 | 56 | ||
| XM_018066274.1 | XP_017921763.1 | 8 - 9 | 2109 | 56 | ||
| XM_018066275.1 | XP_017921764.1 | 7 - 8 | 1968 | 56 | ||
| GenBank mRNA | JO628340.2 | - | 11 - 12 | 2568 | 56 | |
Set Membership: |
|