Search
Search

DIS3L2| Gene Symbol: DIS3L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018058183.1 | XP_017913672.1 | 13 - 14 | 1935 | 93 | |
| XM_018058187.1 | XP_017913676.1 | 13 - 14 | 1861 | 93 | ||
| XM_018058188.1 | XP_017913677.1 | 13 - 14 | 1794 | 93 | ||
| GenBank mRNA | JO432627.2 | - | 5 - 6 | 649 | 93 | |
| JO636071.2 | - | 8 - 9 | 2462 | 93 | ||
| KA305959.1 | - | 13 - 14 | 1791 | 93 | ||
Set Membership: |
|