Search
Search
| Gene Symbol: TEC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005681591.3 | XP_005681648.3 | 6 - 7 | 769 | 69 | |
| XM_005681594.3 | XP_005681651.1 | 6 - 7 | 651 | 69 | ||
| XM_018049427.1 | XP_017904916.1 | 6 - 7 | 770 | 69 | ||
| XM_018049428.1 | XP_017904917.1 | 6 - 7 | 627 | 69 | ||
| XM_018049429.1 | XP_017904918.1 | 6 - 7 | 647 | 69 | ||
Set Membership: |
|