Search
Search
| Gene Symbol: SHPRH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005684887.3 | XP_005684944.1 | 13 - 14 | 3252 | 59 | |
| XM_013966588.2 | XP_013822042.1 | 14 - 15 | 3346 | 59 | ||
| XM_018053308.1 | XP_017908797.1 | 13 - 14 | 3427 | 59 | ||
| XM_018053309.1 | XP_017908798.1 | 13 - 14 | 3252 | 59 | ||
| XM_018053310.1 | XP_017908799.1 | 14 - 15 | 2922 | 59 | ||
| XM_018053311.1 | XP_017908800.1 | 11 - 12 | 2481 | 59 | ||
Set Membership: |
|