Search
Search
| Gene Symbol: SMAD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039538.1 | XP_017895027.1 | 9 - 10 | 1402 | 77 | |
| XM_018039539.1 | XP_017895028.1 | 9 - 10 | 1382 | 77 | ||
| XM_018039540.1 | XP_017895029.1 | 8 - 9 | 1312 | 77 | ||
Set Membership: |
|