Search
Search
| Gene Symbol: NT5M | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064802.1 | XP_017920291.1 | 2 - 3 | 498 | 96 | |
| GenBank mRNA | JO440181.2 | - | 2 - 3 | 311 | 96 | |
| JT676333.1 | - | 4 - 5 | 817 | 96 | ||
Set Membership: |
|