Search
Search

CPT1A| Gene Symbol: CPT1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017978346.1 | XP_017833835.1 | 13 - 14 | 1690 | 61 | |
| XM_017978347.1 | XP_017833836.1 | 13 - 14 | 1885 | 61 | ||
| XM_017978348.1 | XP_017833837.1 | 13 - 14 | 1854 | 61 | ||
| XM_017978349.1 | XP_017833838.1 | 13 - 14 | 1685 | 61 | ||
| XM_017978350.1 | XP_017833839.1 | 12 - 13 | 1607 | 61 | ||
Set Membership: |
|