Search
Search

SOAT1| Gene Symbol: SOAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008984943.2 | XP_008983191.1 | 11 - 12 | 1275 | 96 | |
| XM_008984944.2 | XP_008983192.1 | 12 - 13 | 1303 | 96 | ||
| XM_017966038.1 | XP_017821527.1 | 12 - 13 | 1420 | 96 | ||
| XM_017966039.1 | XP_017821528.1 | 11 - 12 | 1139 | 96 | ||
Set Membership: |
|