Search
Search

HIF1A| Gene Symbol: HIF1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009006132.2 | XP_009004380.1 | 13 - 14 | 2582 | 94 | |
| XM_009006133.2 | XP_009004381.1 | 13 - 14 | 2568 | 94 | ||
| XM_017977146.1 | XP_017832635.1 | 13 - 14 | 4103 | 94 | ||
Set Membership: |
|