Search
Search

RINT1| Gene Symbol: RINT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009002620.2 | XP_009000868.1 | 11 - 12 | 1910 | 102 | |
| XM_017975435.1 | XP_017830924.1 | 11 - 12 | 1671 | 102 | ||
Set Membership: |
|