Search
Search

STK36| Gene Symbol: STK36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008999495.2 | XP_008997743.1 | 3 - 4 | 408 | 68 | |
| XM_017973725.1 | XP_017829214.1 | 3 - 4 | 408 | 68 | ||
| XM_017973726.1 | XP_017829215.1 | 3 - 4 | 408 | 68 | ||
| XM_017973727.1 | XP_017829216.1 | 3 - 4 | 408 | 68 | ||
| XM_017973728.1 | XP_017829217.1 | 3 - 4 | 408 | 68 | ||
| XM_017973729.1 | XP_017829218.1 | 3 - 4 | 408 | 68 | ||
Set Membership: |
|