Search
Search

POMGNT1| Gene Symbol: POMGNT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002750783.4 | XP_002750829.1 | 16 - 17 | 1594 | 57 | |
| XM_009001151.2 | XP_008999399.1 | 16 - 17 | 1594 | 57 | ||
| XM_009001152.1 | XP_008999400.1 | 16 - 17 | 2059 | 57 | ||
| XM_009001153.2 | XP_008999401.1 | 16 - 17 | 1594 | 57 | ||
| XM_017974689.1 | XP_017830178.1 | 15 - 16 | 1363 | 57 | ||
Set Membership: |
|