Search
Search

RSF1| Gene Symbol: RSF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002754792.3 | XP_002754838.1 | 8 - 9 | 2930 | 103 | |
| XM_009007374.2 | XP_009005622.1 | 8 - 9 | 2921 | 103 | ||
Set Membership: |
|