Search
Search

SYT8| Gene Symbol: SYT8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017978381.1 | XP_017833870.1 | 634 | 101 | ||
| XM_017978382.1 | XP_017833871.1 | 631 | 101 | |||
| XM_017978383.1 | XP_017833872.1 | 1230 | 101 | |||
Set Membership: |
|