Search
Search

PRICKLE2| Gene Symbol: PRICKLE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002758524.4 | XP_002758570.1 | 4 - 5 | 8490 | 63 | |
| XM_008981970.2 | XP_008980218.1 | 4 - 5 | 2459 | 63 | ||
| XM_008981972.2 | XP_008980220.1 | 4 - 5 | 679 | 63 | ||
| XM_017965018.1 | XP_017820507.1 | 5 - 6 | 716 | 63 | ||
Set Membership: |
|