Search
Search

LRRC27| Gene Symbol: LRRC27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008978940.1 | XP_008977188.1 | 5 - 6 | 744 | 67 | |
| XM_017963293.1 | XP_017818782.1 | 5 - 6 | 744 | 67 | ||
| XM_017963294.1 | XP_017818783.1 | 4 - 5 | 459 | 67 | ||
Set Membership: |
|