Search
Search

EFTUD2| Gene Symbol: EFTUD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748082.3 | XP_002748128.1 | 25 - 26 | 2868 | 71 | |
| XM_008996800.2 | XP_008995048.1 | 24 - 25 | 2574 | 71 | ||
Set Membership: |
|