Search
Search

SMIM19| Gene Symbol: SMIM19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757007.4 | XP_002757053.1 | 3 - 4 | 402 | 97 | |
| XM_008979407.2 | XP_008977655.1 | 3 - 4 | 310 | 97 | ||
Set Membership: |
|