Search
Search

COL25A1| Gene Symbol: COL25A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008992864.2 | XP_008991112.2 | 25 - 26 | 1561 | 63 | |
| XM_017970350.1 | XP_017825839.1 | 25 - 26 | 1561 | 63 | ||
| XM_017970351.1 | XP_017825840.1 | 24 - 25 | 1537 | 63 | ||
| XM_017970352.1 | XP_017825841.1 | 25 - 26 | 1561 | 63 | ||
| XM_017970353.1 | XP_017825842.1 | 24 - 25 | 1516 | 63 | ||
| XM_017970354.1 | XP_017825843.1 | 24 - 25 | 1516 | 63 | ||
| XM_017970355.1 | XP_017825844.1 | 25 - 26 | 1561 | 63 | ||
| XM_017970356.1 | XP_017825845.1 | 25 - 26 | 1561 | 63 | ||
| XM_017970357.1 | XP_017825846.1 | 25 - 26 | 1561 | 63 | ||
| XM_017970359.1 | XP_017825848.1 | 25 - 26 | 1561 | 63 | ||
| XM_017970360.1 | XP_017825849.1 | 25 - 26 | 1561 | 63 | ||
| XM_017970361.1 | XP_017825850.1 | 25 - 26 | 1561 | 63 | ||
| XM_017970362.1 | XP_017825851.1 | 25 - 26 | 1561 | 63 | ||
Set Membership: |
|