Search
Search

CRYZL1| Gene Symbol: CRYZL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002761400.4 | XP_002761446.1 | 10 - 11 | 1452 | 67 | |
| XM_017967066.1 | XP_017822555.1 | 9 - 10 | 1377 | 67 | ||
Set Membership: |
|