Search
Search

FAM135B| Gene Symbol: FAM135B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002759210.4 | XP_002759256.1 | 5 - 6 | 554 | 60 | |
| XM_008983197.2 | XP_008981445.1 | 5 - 6 | 1428 | 60 | ||
Set Membership: |
|