Search
Search

CASP10| Gene Symbol: CASP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002749624.3 | XP_002749670.1 | 4 - 5 | 989 | 104 | |
| XM_017973640.1 | XP_017829129.1 | 4 - 5 | 989 | 104 | ||
Set Membership: |
|