Search
Search

TRAPPC6A| Gene Symbol: TRAPPC6A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008988219.2 | XP_008986467.1 | 4 - 5 | 393 | 61 | |
| XM_008988221.2 | XP_008986469.1 | 3 - 4 | 329 | 61 | ||
Set Membership: |
|