Search
Search

IKBKG| Gene Symbol: IKBKG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008990116.2 | XP_008988364.1 | 7 - 8 | 1447 | 74 | |
| XM_008990117.2 | XP_008988365.1 | 7 - 8 | 1340 | 74 | ||
| XM_008990118.2 | XP_008988366.1 | 7 - 8 | 1428 | 74 | ||
| XM_008990119.2 | XP_008988367.1 | 7 - 8 | 1446 | 74 | ||
| XM_017968499.1 | XP_017823988.1 | 6 - 7 | 1130 | 74 | ||
Set Membership: |
|