Search
Search

beat-IIIc| Gene Symbol: beat-IIIc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135986.3 | NP_609830.2 | 4 - 5 | 838 | 58 | |
| NM_165209.2 | NP_724042.1 | 4 - 5 | 838 | 58 | ||
| GenBank mRNA | AY075458.1 | - | 4 - 5 | 837 | 58 | |
Set Membership: |
|