Search
Search

M6| Gene Symbol: M6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275237.1 | NP_001262166.1 | 3 - 4 | 823 | 67 | |
| NM_141066.3 | NP_649323.2 | 2 - 3 | 627 | 67 | ||
| NM_168911.2 | NP_730642.1 | 1 - 2 | 398 | 67 | ||
| GenBank mRNA | BT010069.1 | - | 1 - 2 | 398 | 67 | |
| JN872491.1 | - | 2 - 3 | 318 | 67 | ||
Set Membership: |
|