Search
Search

nompA| Gene Symbol: nompA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299383.1 | NP_001286312.1 | 14 - 15 | 2878 | 94 | |
| NM_080092.4 | NP_524831.2 | 15 - 16 | 3631 | 94 | ||
| NM_165821.4 | NP_725030.2 | 15 - 16 | 3631 | 94 | ||
| NM_165822.4 | NP_725031.2 | 14 - 15 | 3502 | 94 | ||
| NM_206096.3 | NP_995818.1 | 14 - 15 | 3502 | 94 | ||
Set Membership: |
|