Search
Search

ADD1| Gene Symbol: ADD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299394.1 | NP_001286323.1 | 2 - 3 | 857 | 64 | |
| NM_136872.3 | NP_610716.1 | 2 - 3 | 857 | 64 | ||
| NM_165856.2 | NP_725094.1 | 2 - 3 | 857 | 64 | ||
| GenBank mRNA | AY051699.1 | - | 2 - 3 | 865 | 64 | |
Set Membership: |
|