Search
Search

psq| Gene Symbol: psq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014520.3 | NP_001014520.2 | 3 - 4 | 1077 | 75 | |
| NM_001316414.1 | NP_001303343.1 | 3 - 4 | 1077 | 75 | ||
| NM_001382049.1 | NP_001369078.1 | 3 - 4 | 1077 | 75 | ||
| NM_078962.4 | NP_523686.2 | 3 - 4 | 1077 | 75 | ||
| NM_165790.3 | NP_724955.1 | 3 - 4 | 1077 | 75 | ||
| NM_206086.3 | NP_995808.1 | 3 - 4 | 920 | 75 | ||
| GenBank mRNA | AY069588.1 | - | 3 - 4 | 998 | 75 | |
Set Membership: |
|