Search
Search

CG6329| Gene Symbol: CG6329 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299464.1 | NP_001286393.1 | 2 - 3 | 444 | 78 | |
| NM_001299465.1 | NP_001286394.1 | 2 - 3 | 361 | 78 | ||
| NM_137060.3 | NP_610904.1 | 2 - 3 | 376 | 78 | ||
| NM_166011.3 | NP_725328.1 | 2 - 3 | 340 | 78 | ||
| NM_166012.3 | NP_725329.1 | 2 - 3 | 686 | 78 | ||
| NM_176169.2 | NP_788349.1 | 2 - 3 | 314 | 78 | ||
| GenBank mRNA | AY069299.1 | - | 2 - 3 | 337 | 78 | |
| BT010209.1 | - | 2 - 3 | 336 | 78 | ||
| KX531333.1 | - | 1 - 2 | 61 | 78 | ||
Set Membership: |
|