Search
Search

bma| Gene Symbol: bma | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104100.2 | NP_001097570.2 | 16 - 17 | 2845 | 86 | |
| NM_001274740.1 | NP_001261669.1 | 16 - 17 | 2890 | 86 | ||
| NM_001382075.1 | NP_001368964.1 | 15 - 16 | 2791 | 86 | ||
| NM_001382076.1 | NP_001369043.1 | 16 - 17 | 2890 | 86 | ||
| GenBank mRNA | BT099519.1 | - | 16 - 17 | 2845 | 86 | |
Set Membership: |
|