Search
Search

Herp| Gene Symbol: Herp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135307.3 | NP_609151.2 | 3 - 4 | 1546 | 70 | |
| NM_164772.2 | NP_723316.1 | 3 - 4 | 1335 | 70 | ||
| GenBank mRNA | BT012479.1 | - | 3 - 4 | 1305 | 70 | |
Set Membership: |
|