Search
Search

Hnf4| Gene Symbol: Hnf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103656.3 | NP_001097126.1 | 3 - 4 | 1995 | 64 | |
| NM_001298829.1 | NP_001285758.1 | 3 - 4 | 1186 | 64 | ||
| NM_057539.5 | NP_476887.2 | 4 - 5 | 1675 | 64 | ||
| NM_164833.2 | NP_723413.1 | 3 - 4 | 1134 | 64 | ||
| NM_164834.2 | NP_723414.2 | 4 - 5 | 2244 | 64 | ||
| GenBank mRNA | BT003533.1 | - | 4 - 5 | 1671 | 64 | |
Set Membership: |
|