Search
Search

Snx6| Gene Symbol: Snx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259010.2 | NP_001245939.1 | 7 - 8 | 1389 | 61 | |
| NM_135355.4 | NP_609199.1 | 6 - 7 | 1371 | 61 | ||
| GenBank mRNA | AY069492.1 | - | 6 - 7 | 1343 | 61 | |
Set Membership: |
|