Search
Search

nej| Gene Symbol: nej | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201646.2 | NP_001188575.1 | 9 - 10 | 8271 | 62 | |
| NM_001201647.2 | NP_001188576.1 | 9 - 10 | 8223 | 62 | ||
| NM_001272458.1 | NP_001259387.1 | 9 - 10 | 8125 | 62 | ||
| NM_079903.4 | NP_524642.2 | 9 - 10 | 8300 | 62 | ||
Set Membership: |
|