Search
Search

ND-ASHI| Gene Symbol: ND-ASHI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031877.2 | NP_001027048.1 | 1 - 2 | 263 | 77 | |
| NM_001298022.1 | NP_001284951.1 | 1 - 2 | 164 | 77 | ||
| NM_132110.2 | NP_572338.1 | 2 - 3 | 779 | 77 | ||
| GenBank mRNA | BT120041.1 | - | 2 - 3 | 778 | 77 | |
Set Membership: |
|