Search
Search

Uch| Gene Symbol: Uch | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201752.2 | NP_001188681.1 | 3 - 4 | 451 | 64 | |
| NM_001272986.1 | NP_001259915.1 | 3 - 4 | 442 | 64 | ||
| NM_057592.4 | NP_476940.1 | 3 - 4 | 603 | 64 | ||
| GenBank mRNA | AF145600.1 | - | 3 - 4 | 427 | 64 | |
Set Membership: |
|