Search
Search

Shaw| Gene Symbol: Shaw | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144310.1 | NP_001137782.1 | 6 - 7 | 1514 | 74 | |
| NM_164554.3 | NP_722937.2 | 9 - 10 | 1926 | 74 | ||
| GenBank mRNA | AY071475.1 | - | 9 - 10 | 1924 | 74 | |
Set Membership: |
|